Thermo Scientific Chemicals

Nigrosine, pure, water soluble, high purity biological stain

Catalog number: 189480250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Nigrosine, pure, water soluble, high purity biological stain

Catalog number: 189480250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 189480250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
8005-03-6
IUPAC Name
disodium (3E)-5-amino-6-[(1E)-2-(4-nitrophenyl)diazen-1-yl]-4-oxo-3-(2-phenylhydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate
Molecular Formula
C22H14N6Na2O9S2
InChI Key
HKBVRFLHNUEVRO-HMDKGIQMSA-L
SMILES
[Na+].[Na+].NC1=C2C(=O)\C(=N/NC3=CC=CC=C3)C(=CC2=CC(=C1\N=N\C1=CC=C(C=C1)[N+]([O-])=O)S([O-])(=O)=O)S([O-])(=O)=O
Appearance (Color)
Black
Appearance (Form)
Crystalline powder or crystals
UV spectrum
Complies
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text