Thermo Scientific Chemicals

Cesium chloride, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 189500010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Cesium chloride, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 189500010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 189500010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7647-17-8
IUPAC Name
caesium(1+) chloride
Molecular Formula
ClCs
InChI Key
AIYUHDOJVYHVIT-UHFFFAOYSA-M
SMILES
[Cl-].[Cs+]
Appearance (Form)
Powder or crystals
Silicon dioxide (SiO2)
=<20 ppm
Sodium (Na)
=<50 ppm
Rubidium (Rb)
=<200 ppm
Calcium (Ca)
=<10 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text