Thermo Scientific Chemicals

Diethyl bis(hydroxymethyl)malonate, 95%, Thermo Scientific Chemicals

Catalog number: 189530250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Diethyl bis(hydroxymethyl)malonate, 95%, Thermo Scientific Chemicals

Catalog number: 189530250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 189530250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
20605-01-0
IUPAC Name
1,3-diethyl 2,2-bis(hydroxymethyl)propanedioate
Molecular Formula
C9H16O6
InChI Key
WIOHBOKEUIHYIC-UHFFFAOYSA-N
SMILES
CCOC(=O)C(CO)(CO)C(=O)OCC
Appearance (Color)
White to off-white
Melting point
45°C to 52°C
Appearance (Form)
Crystalline solid
Infrared spectrum
Conforms
GC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text