Diethyl bis(hydroxymethyl)malonate, 95%, Thermo Scientific Chemicals
Diethyl bis(hydroxymethyl)malonate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl bis(hydroxymethyl)malonate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
View all
Glass bottle
Catalog number 189531000
Price (USD)/ Each
221.00
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
221.00
Add to cart
Diethyl bis(hydroxymethyl)malonate, 95%, Thermo Scientific Chemicals
Catalog number189531000
Price (USD)/ Each
221.00
-
Add to cart
Chemical Identifiers
CAS20605-01-0
IUPAC Name1,3-diethyl 2,2-bis(hydroxymethyl)propanedioate
Molecular FormulaC9H16O6
InChI KeyWIOHBOKEUIHYIC-UHFFFAOYSA-N
SMILESCCOC(=O)C(CO)(CO)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Melting point45°C to 52°C
Appearance (Form)Crystalline solid
Infrared spectrumConforms
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only