Thermo Scientific™
Cryo Gloves - Elbow Length
Catalog number: 189566
Have Questions?
Thermo Scientific™
Cryo Gloves - Elbow Length
Catalog number: 189566
Have Questions?
Safely handle samples from ultra-low and cryogenic storage with these cryo gloves which offer a high level of thermal protection.
Catalog Number
189566
Unit Size
Each
Size
X-Large
Price (USD)
Full specifications
DescriptionElbow Length
ColorBlue
HandSpecific
SizeX-Large
For Use With (Application)Handling Samples from Ultra Low Temperature and Cryogenic Storage
Length (English)18 in.
Length (Metric)45.7 cm
Unit SizeEach
Catalog Number
Unit Size
Each
Size
Medium
Price (USD)
Full specifications
DescriptionElbow Length
ColorBlue
HandSpecific
SizeMedium
For Use With (Application)Handling Samples from Ultra Low Temperature and Cryogenic Storage
Length (English)18 in.
Length (Metric)45.7 cm
Unit SizeEach
Catalog Number
Unit Size
Each
Size
Large
Price (USD)
Full specifications
DescriptionElbow Length
ColorBlue
HandSpecific
SizeLarge
For Use With (Application)Handling Samples from Ultra Low Temperature and Cryogenic Storage
Length (English)18 in.
Length (Metric)45.7 cm
Unit SizeEach
Showing 3 of 3
Catalog NumberSpecificationsUnit SizeSizePrice (USD)
189566Full specifications
EachX-LargeRequest A Quote
DescriptionElbow Length
ColorBlue
HandSpecific
SizeX-Large
For Use With (Application)Handling Samples from Ultra Low Temperature and Cryogenic Storage
Length (English)18 in.
Length (Metric)45.7 cm
Unit SizeEach
189564Full specifications
EachMediumRequest A Quote
DescriptionElbow Length
ColorBlue
HandSpecific
SizeMedium
For Use With (Application)Handling Samples from Ultra Low Temperature and Cryogenic Storage
Length (English)18 in.
Length (Metric)45.7 cm
Unit SizeEach
189565Full specifications
EachLargeRequest A Quote
DescriptionElbow Length
ColorBlue
HandSpecific
SizeLarge
For Use With (Application)Handling Samples from Ultra Low Temperature and Cryogenic Storage
Length (English)18 in.
Length (Metric)45.7 cm
Unit SizeEach
Showing 3 of 3
  • Comfortable, lightweight, and breathable protection
  • Side release buckles for quick on/off
  • Available in a variety of sizes