Thermo Scientific Chemicals

4-Methyl-2-oxopentanoic acid, sodium salt, hydrate, 98+%, Thermo Scientific Chemicals

Catalog number: 189710010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Methyl-2-oxopentanoic acid, sodium salt, hydrate, 98+%, Thermo Scientific Chemicals

Catalog number: 189710010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 189710010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
332360-07-3
IUPAC Name
4-methyl-2-oxopentanoate
Molecular Formula
C6H9O3
InChI Key
BKAJNAXTPSGJCU-UHFFFAOYSA-M
SMILES
CC(C)CC(=O)C([O-])=O
Appearance (Form)
Powder
Titration with HClO4
>=98.0 % (On dry substance)
Infrared spectrum
Conforms
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text