3-Methyl-2-oxobutanoic acid, sodium salt, 98+%, Thermo Scientific Chemicals
3-Methyl-2-oxobutanoic acid, sodium salt, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methyl-2-oxobutanoic acid, sodium salt, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 189720010
Price (USD)/ Each
65.65
Online exclusive
72.90 
Save 7.25 (10%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
65.65
Online exclusive
72.90 
Save 7.25 (10%)
Add to cart
3-Methyl-2-oxobutanoic acid, sodium salt, 98+%, Thermo Scientific Chemicals
Catalog number189720010
Price (USD)/ Each
65.65
Online exclusive
72.90 
Save 7.25 (10%)
-
Add to cart
Chemical Identifiers
CAS3715-29-5
IUPAC Namesodium 3-methyl-2-oxobutanoate
Molecular FormulaC5H7NaO3
InChI KeyWIQBZDCJCRFGKA-UHFFFAOYSA-M
SMILES[Na+].CC(C)C(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO498.0 to 102.0 %
Appearance (Color)White to light yellow
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
Melting point227°C to 231°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only