Thermo Scientific Chemicals

Sodium hydride, 60% dispersion in mineral oil, in soluble bags, Thermo Scientific Chemicals

Catalog number: 189862500
250 g, Each
Thermo Scientific Chemicals

Sodium hydride, 60% dispersion in mineral oil, in soluble bags, Thermo Scientific Chemicals

Catalog number: 189862500
250 g, Each
Quantity
Catalog number: 189862500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7646-69-7
IUPAC Name
sodium
Molecular Formula
Na
InChI Key
KEAYESYHFKHZAL-UHFFFAOYSA-N
SMILES
[Na]
Appearance (Color)
White to grey
Appearance (Form)
Tacky powder
Assay
57 to 63 % (NaH, Gasvolumetric)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text