1x1 image pixel for data collection
L(+)-Lactic acid, 90% solution in water
L(+)-Lactic acid, 90% solution in water
L(+)-Lactic acid, 90% solution in water
L(+)-Lactic acid, 90% solution in water
Thermo Scientific Chemicals

L(+)-Lactic acid, 90% solution in water

CAS: 79-33-4 | C3H6O3 | 90.078 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
5 L
Packaging:
View all
Glass bottle
Catalog number 189872500
Price (USD)
56.65
Special offer
Online exclusive
Ends: 26-Sep-2025
69.60
Save 12.95 (19%)
Each
-
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
56.65
Special offer
Online exclusive
Ends: 26-Sep-2025
69.60
Save 12.95 (19%)
Each
Add to cart
Chemical Identifiers
CAS79-33-4
IUPAC Name(2S)-2-hydroxypropanoic acid
Molecular FormulaC3H6O3
InChI KeyJVTAAEKCZFNVCJ-REOHCLBHSA-N
SMILESC[C@H](O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Titration excess NaOH>=88 %
Density(20°C) 1.2 to 1.21 g/ml
IdentificationPasses test
Infrared spectrumConforms
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0474222Certificate of AnalysisJul 09, 2025189870051, 189872500, 189870010
A0471892Certificate of AnalysisMay 17, 2025189870051, 189872500, 189870010
A0460875Certificate of AnalysisJun 11, 2024189870051, 189872500, 189870010
A0463441Certificate of AnalysisJun 06, 2024189870051, 189872500, 189870010
A0449919Certificate of AnalysisFeb 03, 2023189870051, 189872500, 189870010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection