Thermo Scientific Chemicals

Tetrabromophenol Blue, pure

Catalog number: 189910250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetrabromophenol Blue, pure

Catalog number: 189910250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 189910250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4430-25-5
IUPAC Name
4,5,6,7-tetrabromo-3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular Formula
C19H6Br8O5S
InChI Key
QPMIVFWZGPTDPN-UHFFFAOYSA-N
SMILES
OC1=C(Br)C=C(C=C1Br)C1(OS(=O)(=O)C2=C(Br)C(Br)=C(Br)C(Br)=C12)C1=CC(Br)=C(O)C(Br)=C1
Appearance (Form)
Powder
UV spectrum
Complies
UV Lambda max
608 to 618 nm
Specific extinction E (1%/1cm)
(0.01 g/l 50% ethanol, buffer pH 5)
Appearance (Color)
Beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text