Light /UV Protection for Vertical Light Chambers
Product Image
Thermo Scientific™
Light /UV Protection for Vertical Light Chambers
Protection for operator, light switches off automatically when door is opened
Have Questions?
Catalog NumberFor Use With (Equipment)
1900663TSAll Thermo Scientific and Forma vertical light chambers - models 3906, 3909, 3928, 3942, 3943
Catalog number 1900663TS
Price (KRW)
-
Request A Quote
For Use With (Equipment):
All Thermo Scientific and Forma vertical light chambers - models 3906, 3909, 3928, 3942, 3943
Ask our AI about this Product
Light / UV protection is a feature to protect the operator's eyes. This factory-installed option will switch off the vertical light as the door is opened. Recommended for ICH photostability testing. Not recommended for plant growth / animal hatching, sinc
  • Light / UV protection for operator's eyes.
  • Factory-installed option will switch off the vertical light as the door is opened
  • Recommended for ICH photo stability testing
  • Not recommended for plant growth/animal hatching since switching light on and off could stress plants and animals
  • Operator can wear goggles as an alternative protection.
Specifications
DescriptionLight / UV protection - light off with open door
For Use With (Application)Light application - recommended for ICH photostability testing
For Use With (Equipment)All Thermo Scientific and Forma vertical light chambers - models 3906, 3909, 3928, 3942, 3943
IncludesFactory installed door switch
LightingLED
TypeAccessory, Light/UV Protection
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection