Thermo Scientific Chemicals

α-Naphtholphthalein, pure, indicator

Catalog number: 190120050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

α-Naphtholphthalein, pure, indicator

Catalog number: 190120050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 190120050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
596-01-0
IUPAC Name
3,3-bis(4-hydroxynaphthalen-1-yl)-1,3-dihydro-2-benzofuran-1-one
Molecular Formula
C28H18O4
InChI Key
HQHBAGKIEAOSNM-UHFFFAOYSA-N
SMILES
OC1=C2C=CC=CC2=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=C2C=CC=CC2=C(O)C=C1
Appearance (Form)
Powder
Loss on drying
=<5 % (110°C, 1 h)
Specific absorbance A (1%/1cm)
>=550 (at lamda max)
Appearance (Color)
Beige to pink to brown
UV Lambda max
648 to 654 nm (in 0.1N NaOH)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text