Sudan Black B, high purity biological stain
Sudan Black B, high purity biological stain
Sudan Black B, high purity biological stain
Sudan Black B, high purity biological stain
Thermo Scientific Chemicals

Sudan Black B, high purity biological stain

CAS: 4197-25-5 | C29H24N6 | 456.553 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 190160250
Price (USD)
72.65
Special offer
Online exclusive
Ends: 26-Sep-2025
89.30
Save 16.65 (19%)
Each
In stock
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
72.65
Special offer
Online exclusive
Ends: 26-Sep-2025
89.30
Save 16.65 (19%)
Each
Add to cart
Chemical Identifiers
CAS4197-25-5
IUPAC Name2,2-dimethyl-6-{2-[4-(2-phenyldiazen-1-yl)naphthalen-1-yl]diazen-1-yl}-2,3-dihydro-1H-perimidine
Molecular FormulaC29H24N6
InChI KeyYCUVUDODLRLVIC-UHFFFAOYSA-N
SMILESCC1(C)NC2=CC=C(N=NC3=C4C=CC=CC4=C(C=C3)N=NC3=CC=CC=C3)C3=CC=CC(N1)=C23
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark brown to black
Infrared spectrumConforms
Appearance (Form)Crystalline powder
UV spectrumComplies
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection