Sudan Black B, high purity biological stain
Sudan Black B, high purity biological stain
Sudan Black B, high purity biological stain
Sudan Black B, high purity biological stain
Thermo Scientific Chemicals

Sudan Black B, high purity biological stain

CAS: 4197-25-5 | C29H24N6 | 456.553 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 190161000
Price (USD)
207.65
Online Exclusive
230.00
Save 22.35 (10%)
Each
In stock
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
207.65
Online Exclusive
230.00
Save 22.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS4197-25-5
IUPAC Name2,2-dimethyl-6-{2-[4-(2-phenyldiazen-1-yl)naphthalen-1-yl]diazen-1-yl}-2,3-dihydro-1H-perimidine
Molecular FormulaC29H24N6
InChI KeyYCUVUDODLRLVIC-UHFFFAOYSA-N
SMILESCC1(C)NC2=CC=C(N=NC3=C4C=CC=CC4=C(C=C3)N=NC3=CC=CC=C3)C3=CC=CC(N1)=C23
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark brown to black
Infrared spectrumConforms
Appearance (Form)Crystalline powder
UV spectrumComplies
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0468328Certificate of AnalysisJan 31, 2025190161000, 190160250
A0459036Certificate of AnalysisJan 03, 2024190161000, 190160250
A0454982Certificate of AnalysisAug 25, 2023190161000, 190160250
A0395098Certificate of AnalysisDec 15, 2022190161000, 190160250
A0442762Certificate of AnalysisJun 10, 2022190161000, 190160250
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection