Thermo Scientific Chemicals

Tropaeolin O, pure

Catalog number: 190180250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Tropaeolin O, pure

Catalog number: 190180250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 190180250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
547-57-9
IUPAC Name
4-[2-(2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene)hydrazin-1-yl]benzene-1-sulfonic acid sodium
Molecular Formula
C12H10N2NaO5S
InChI Key
YOGUPPQYRWYKGT-UHFFFAOYSA-N
SMILES
[Na].OC1=CC(=O)C=CC1=NNC1=CC=C(C=C1)S(O)(=O)=O
Appearance (Color)
Rust brown to dark brown
Infrared spectrum
Conforms
Visual transition interval
from pH 11.1 (yellow) to pH 12.7 (orange)
Appearance (Form)
Powder
Solubility
(in water) Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text