Chemical Identifiers
CAS547-57-9
IUPAC Name4-[2-(2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene)hydrazin-1-yl]benzene-1-sulfonic acid sodium
Molecular FormulaC12H10N2NaO5S
InChI KeyYOGUPPQYRWYKGT-UHFFFAOYSA-N
SMILES[Na].OC1=CC(=O)C=CC1=NNC1=CC=C(C=C1)S(O)(=O)=O
View more