Thermo Scientific Chemicals

Carmine, High Purity Biological Stain (Alum Lake of Carminic Acid)

Catalog number: 190200250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Carmine, High Purity Biological Stain (Alum Lake of Carminic Acid)

Catalog number: 190200250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 190200250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1390-65-4
IUPAC Name
3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracene-2-carboxylic acid
Molecular Formula
C22H20O13
InChI Key
DGQLVPJVXFOQEV-UHFFFAOYNA-N
SMILES
CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O
Appearance (Color)
Dark red to purple
Assay
>=42.0 % (Carminic acid)
Ash
=<12 %
Infrared spectrum
Conforms
Loss on drying
=<20 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text