Thermo Scientific Chemicals

Litmus, pure, indicator grade

Catalog number: 190220250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Litmus, pure, indicator grade

Catalog number: 190220250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 190220250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1393-92-6
Solubility
(0.01 % in water) Opaque dark blue
Appearance (Color)
Dark blue to black
Infrared spectrum
Conforms
Visual transition interval
from pH 4.8 (red) to pH 8.3 (blue)
Appearance (Form)
Powder or granules

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text