Litmus, pure, indicator grade
Litmus, pure, indicator grade
Thermo Scientific Chemicals

Litmus, pure, indicator grade

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 190225000
Price (USD)/ Each
436.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
514.00 
Save 77.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
436.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
514.00 
Save 77.35 (15%)
Add to cart
Litmus, pure, indicator grade
Catalog number190225000
Price (USD)/ Each
436.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
514.00 
Save 77.35 (15%)
-
Add to cart
Chemical Identifiers
CAS1393-92-6
SpecificationsSpecification SheetSpecification Sheet
Solubility(0.01 % in water) Opaque dark blue
Appearance (Color)Dark blue to black
Infrared spectrumConforms
Visual transition intervalfrom pH 4.8 (red) to pH 8.3 (blue)
Appearance (Form)Powder or granules
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only