1x1 image pixel for data collection
Sodium azide, 99%, extra pure
Sodium azide, 99%, extra pure
Sodium azide, 99%, extra pure
Sodium azide, 99%, extra pure
Thermo Scientific Chemicals

Sodium azide, 99%, extra pure

CAS: 26628-22-8 | N3Na | 65.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Catalog number 190380050
Price (USD)
19.65
Special offer
Online exclusive
Ends: 26-Sep-2025
24.00
Save 4.35 (18%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
19.65
Special offer
Online exclusive
Ends: 26-Sep-2025
24.00
Save 4.35 (18%)
Each
Add to cart
Chemical Identifiers
CAS26628-22-8
IUPAC Namesodium azide
Molecular FormulaN3Na
InChI KeyPXIPVTKHYLBLMZ-UHFFFAOYSA-N
SMILES[Na+].[N-]=[N+]=[N-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Insoluble matter=<0.05 % (in water)
Sodium carbonate (Na2CO3)=<0.15 %
Appearance (Color)White
Loss on drying=<0.5 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, For Azidation/Diazo transfer
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection