Thermo Scientific Chemicals

Sodium azide, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 190381000
100 g, Each
Thermo Scientific Chemicals

Sodium azide, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 190381000
100 g, Each
Quantity
Catalog number: 190381000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
26628-22-8
IUPAC Name
sodium azide
Molecular Formula
N3Na
InChI Key
PXIPVTKHYLBLMZ-UHFFFAOYSA-N
SMILES
[Na+].[N-]=[N+]=[N-]
Appearance (Form)
Crystalline powder
Insoluble matter
=<0.05 % (in water)
Sodium carbonate (Na2CO3)
=<0.15 %
Appearance (Color)
White
Loss on drying
=<0.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, For Azidation/Diazo transfer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text