Thermo Scientific Chemicals

Molybdenum hexacarbonyl, 98%, Thermo Scientific Chemicals

Catalog number: 190390500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Molybdenum hexacarbonyl, 98%, Thermo Scientific Chemicals

Catalog number: 190390500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 190390500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13939-06-5
IUPAC Name
hexakis(methanidylidyneoxidanium) molybdenum
Molecular Formula
C6MoO6
InChI Key
KMKBZNSIJQWHJA-UHFFFAOYSA-N
SMILES
[Mo].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
Infrared spectrum
Conforms
Elemental Analysis
>=97.5 %
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text