Tris(triphenylphosphine)rhodium(I) chloride, 98%, Thermo Scientific Chemicals
Tris(triphenylphosphine)rhodium(I) chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tris(triphenylphosphine)rhodium(I) chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 190420010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS14694-95-2
IUPAC Nameλ¹-rhodium(1+) tris(triphenylphosphane) chloride
Molecular FormulaC54H45ClP3Rh
InChI KeyIXAYKDDZKIZSPV-UHFFFAOYSA-M
SMILES[Cl-].[Rh+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red-brown to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Gravimetry10.8 to 11.3 % (Rh)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only