Thermo Scientific Chemicals

Chromium(III) chloride, 99%, anhydrous, Thermo Scientific Chemicals

Catalog number: 190470250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Chromium(III) chloride, 99%, anhydrous, Thermo Scientific Chemicals

Catalog number: 190470250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 190470250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10025-73-7
IUPAC Name
chromium(3+) hexahydrate trichloride
Molecular Formula
Cl3CrH12O6
InChI Key
LJAOOBNHPFKCDR-UHFFFAOYSA-K
SMILES
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]
Appearance (Color)
Purple
Appearance (Form)
Glistening powder or flakes
Assay
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text