Thermo Scientific Chemicals

Mercury(II) bromide, 99+%, Thermo Scientific Chemicals

Catalog number: 190491000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Mercury(II) bromide, 99+%, Thermo Scientific Chemicals

Catalog number: 190491000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 190491000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7789-47-1
IUPAC Name
mercury(2+) dibromide
Molecular Formula
Br2Hg
InChI Key
NGYIMTKLQULBOO-UHFFFAOYSA-L
SMILES
[Br-].[Br-].[Hg++]
Appearance (Color)
White
Insoluble matter
=<0.05 % (in methanol)
Appearance (Form)
Crystalline powder
Chloride (Cl)
=<2000 ppm
Assay
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text