Chemical Identifiers
CAS5423-07-4
IUPAC Namedisodium 3-[(1E)-2-(2,4-dinitrophenyl)diazen-1-yl]-4-hydroxynaphthalene-2,7-disulfonate
Molecular FormulaC16H8N4Na2O11S2
InChI KeyXBLIFEQTVVSTIM-QIKYXUGXSA-L
SMILES[Na+].[Na+].OC1=C2C=CC(=CC2=CC(=C1N=NC1=C(C=C(C=C1)[N+]([O-])=O)[N+]([O-])=O)S([O-])(=O)=O)S([O-])(=O)=O
View more