Poly(methyl methacrylate), ∽ M.W. 15000 (GPC), Thermo Scientific Chemicals
Poly(methyl methacrylate), ∽ M.W. 15000 (GPC), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(methyl methacrylate), ∽ M.W. 15000 (GPC), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
50 g
Packaging:
Plastic bottle
Catalog number 190691000
Price (USD)/ Each
170.65
Online exclusive
189.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
170.65
Online exclusive
189.00 
Save 18.35 (10%)
Add to cart
Poly(methyl methacrylate), ∽ M.W. 15000 (GPC), Thermo Scientific Chemicals
Catalog number190691000
Price (USD)/ Each
170.65
Online exclusive
189.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS9011-14-7
Molecular Formula(C5H8O2)n
SMILESCOC(=O)C(C)(-*)C-*
MDL NumberMFCD00134349
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Beads
Loss on drying=<3 %
Infrared spectrumConforms
Appearance (Color)White
Additional infobead size (typically): 10 to 100 micron
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only