iConnect™ Split/Splitless (SSL) Injector Module
iConnect™ Split/Splitless (SSL) Injector Module
iConnect™ Split/Splitless (SSL) Injector Module
iConnect™ Split/Splitless (SSL) Injector Module
Thermo Scientific™
iConnect™ Split/Splitless (SSL) Injector Module
Employ the optimized thermal profile of these iConnect™ Split/Splitless (SSL) GC Injectors to avoid sample discrimination and provide accurate injection of a broader range of analytes.
Have Questions?
Catalog NumberDescriptionType
19070010iConnect Split/Splitless (SSL) Injector moduleiConnect SSL Injector
Catalog number 19070010
Price (USD)
4,820.00
Each
-
Add to cart
Description:
iConnect Split/Splitless (SSL) Injector module
Type:
iConnect SSL Injector
Price (USD)
4,820.00
Each
Add to cart
Ask our AI about this Product
Gain greater versatility in gas chromatography with instant connect (iConnect™) injectors for Thermo Scientific™ TRACE™ 1300 and 1600 Series GC systems. Thermo Scientific™ iConnect Split/Splitless (SSL) Injectors have an optimized thermal profile that helps prevent sample discrimination in split and splitless mode and accurately injects a broader range of analytes than other GC injector modules. Their cool injector head exerts minimum septum thermal stress to reduce bleed and extend septa lifetime. Flexible injector configuration enables quick and easy implementation of existing and validated methods to maximize lab productivity.
FOR US ONLINE SALES ONLY
  • Suitable for all capillary columns (50 μm to 530 μm inner diameter)
  • Supports concurrent solvent recondensation (CSR) for large volume injections (up to 50 μL)
  • Compatible with 1/8 inch and 1/16 inch packed column using adapters
  • Supports P&T/TD/HS by special adapter
  • Compatible with the Merlin Microseal™ septum
  • Maximum temperature: 400°C
SSL injector with integrated concurrent backflush
  • Reverse the flow inside the injector
  • Eliminate heavy or “undesired” compounds concurrently during the analysis run
  • Increase throughput
SSL injector advantages
  • Perform reproducible, accurate, and robust sample injection
  • Avoid sample discrimination and accurately inject a broader range of analytes
  • Reduce bleed and extend septa lifetime
  • Implement existing and validated methods quickly and easily
  • Use many standard consumables such as liners, septa, and ferrules already in your lab
  • Gain easy and immediate access to the septum and liner for simple and quick maintenance without tools or removing gas lines thanks to TRACE 1300 and 1600 Series iConnect modules
  • Extract and restore the injector body quickly for cleaning when needed, such as after difficult, sticky samples
Reduce downtime, simplify maintenance, and enhance GC performance
iConnect injectors and detectors redefine GC and GC-MS flexibility and usability. In two minutes, without training or tools, you can change these modules to:
  • Reconfigure your system for a specific workflow or method
  • Perform maintenance and dramatically reduce maintenance downtime
  • Switch injectors and detectors between TRACE GC systems
Specifications
DescriptioniConnect Split/Splitless (SSL) Injector module
QuantityEach
For Use With (Equipment)TRACE 1300 and 1600 Series Gas Chromatographs; AI/AS 1310, AI/AS 1610, TriPlus 100 LS, TriPlus RSH, TriPlus RSH SMART, and TriPlus 500 HS Autosamplers
Injection ModeSplit/Splitless
TypeiConnect SSL Injector
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection