Poly(caprolactone) triol, average M.W. 300, Thermo Scientific Chemicals
Poly(caprolactone) triol, average M.W. 300, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(caprolactone) triol, average M.W. 300, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Packaging:
Glass bottle
Catalog number 190730250
Price (USD)/ Each
66.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
77.70 
Save 11.05 (14%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
66.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
77.70 
Save 11.05 (14%)
Add to cart
Poly(caprolactone) triol, average M.W. 300, Thermo Scientific Chemicals
Catalog number190730250
Price (USD)/ Each
66.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
77.70 
Save 11.05 (14%)
-
Add to cart
Chemical Identifiers
CAS37625-56-2
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<2 %
Appearance (Color)Clear colorless
Appearance (Form)Viscous liquid
Infrared spectrumConforms
Refractive index1.4750 to 1.4780 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only