Ethyl cellulose, ethoxyl content 48%, 10 cps
Ethyl cellulose, ethoxyl content 48%, 10 cps
Ethyl cellulose, ethoxyl content 48%, 10 cps
Ethyl cellulose, ethoxyl content 48%, 10 cps
Thermo Scientific Chemicals

Ethyl cellulose, ethoxyl content 48%, 10 cps

CAS: 9004-57-3 | C23H24N6O4 | 448.483 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
Packaging:
Glass bottle
Catalog number 190965000
Price (USD)
156.65
Special offer
Online exclusive
184.00
Save 27.35 (15%)
Each
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
156.65
Special offer
Online exclusive
184.00
Save 27.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS9004-57-3
IUPAC Name2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate
Molecular FormulaC23H24N6O4
InChI KeyARSKJXYLLONUAJ-UHFFFAOYSA-N
SMILESCCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Viscosity8 to 12 cP (5% in 80/20 toluene/ethanol, 25°C)
Appearance (Form)Powder or granules
Assay47.5 to 49.5 % (Ethoxyl content)
Appearance (Color)White to yellow
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0468355Certificate of AnalysisJan 31, 2025190962500, 190965000
A0469124Certificate of AnalysisJan 31, 2025190962500, 190965000
A0457666Certificate of AnalysisNov 21, 2023190962500, 190965000
A0447572Certificate of AnalysisJan 05, 2023190962500, 190965000
A0400529Certificate of AnalysisDec 15, 2022190962500, 190965000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection