Thermo Scientific Chemicals

Ethyl cellulose, ethoxyl content 48%, 22cps

Catalog number: 190972500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl cellulose, ethoxyl content 48%, 22cps

Catalog number: 190972500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 190972500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9004-57-3
IUPAC Name
2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate
Molecular Formula
C23H24N6O4
InChI Key
ARSKJXYLLONUAJ-UHFFFAOYSA-N
SMILES
CCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N
Infrared spectrum
Conforms
Viscosity
18 to 24 cP (5% soln. in 80/20 toluene/ethanol, 25°C)
Appearance (Form)
Crystalline powder or granules
Assay
48 to 49.5 % (Ethoxyl)
Appearance (Color)
White to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text