Thermo Scientific Chemicals

Potassium hexafluorophosphate, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 191090500
50 g, Plastic bottle, Each
Thermo Scientific Chemicals

Potassium hexafluorophosphate, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 191090500
50 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 191090500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17084-13-8
IUPAC Name
potassium hexafluoro-λ⁵-phosphanuide
Molecular Formula
F6KP
InChI Key
YZDGRYDIGCWVND-UHFFFAOYSA-N
SMILES
[K+].F[P-](F)(F)(F)(F)F
Assay
>=98.5 %
Loss on drying
=<0.1 % (100°C)
Appearance (Form)
Crystalline powder
Free fluoride
=<0.5 %
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text