Potassium hexafluorophosphate, 99%, extra pure, Thermo Scientific Chemicals
Potassium hexafluorophosphate, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium hexafluorophosphate, 99%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 191092500
Price (USD)/ Each
101.65
Online exclusive
119.00 
Save 17.35 (15%)
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
101.65
Online exclusive
119.00 
Save 17.35 (15%)
Add to cart
Potassium hexafluorophosphate, 99%, extra pure, Thermo Scientific Chemicals
Catalog number191092500
Price (USD)/ Each
101.65
Online exclusive
119.00 
Save 17.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namepotassium hexafluoro-λ⁵-phosphanuide
Molecular FormulaF6KP
InChI KeyYZDGRYDIGCWVND-UHFFFAOYSA-N
SMILES[K+].F[P-](F)(F)(F)(F)F
Molecular Weight (g/mol)184.06
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=98.5 %
Loss on drying=<0.1 % (100°C)
Appearance (Form)Crystalline powder
Free fluoride=<0.5 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only