Hexafluorophosphoric acid, 60 wt.% solution in water, Thermo Scientific Chemicals
Hexafluorophosphoric acid, 60 wt.% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hexafluorophosphoric acid, 60 wt.% solution in water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
Packaging:
Plastic bottle
Catalog number 191115000
Price (USD)/ Each
93.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
110.00 
Save 16.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
93.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
110.00 
Save 16.35 (15%)
Add to cart
Hexafluorophosphoric acid, 60 wt.% solution in water, Thermo Scientific Chemicals
Catalog number191115000
Price (USD)/ Each
93.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
110.00 
Save 16.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namehydrogen hexafluoro-λ⁵-phosphanuide
Molecular FormulaF6HP
InChI KeyLJQLCJWAZJINEB-UHFFFAOYSA-O
SMILES[H+].F[P-](F)(F)(F)(F)F
Molecular Weight (g/mol)145.97
View more
SpecificationsSpecification SheetSpecification Sheet
Assay Titrimetry>=55.0 %
Appearance (Color)Colorless to yellow-green
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only