Thermo Scientific Chemicals

Osmium tetroxide, 99.9+%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 191182500
250 mg, Each
Thermo Scientific Chemicals

Osmium tetroxide, 99.9+%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 191182500
250 mg, Each
Quantity
Catalog number: 191182500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20816-12-0
IUPAC Name
tetraoxoosmium
Molecular Formula
O4Os
InChI Key
VUVGYHUDAICLFK-UHFFFAOYSA-N
SMILES
O=[Os](=O)(=O)=O
Osmium (Os)
74.2 to 75.4 %
Solubility
Passes test
Appearance (Color)
Yellow
Appearance (Form)
Crystals
Non-volatile matter
=<0.2 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis, Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text