Osmium tetroxide, 99.9+%, (trace metal basis), Thermo Scientific Chemicals
Osmium tetroxide, 99.9+%, (trace metal basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Osmium tetroxide, 99.9+%, (trace metal basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
100 mg
250 mg
1 g
5 g
Catalog number 191185000
Price (USD)/ Each
135.65
Online exclusive
150.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)/ Each
135.65
Online exclusive
150.00 
Save 14.35 (10%)
Add to cart
Osmium tetroxide, 99.9+%, (trace metal basis), Thermo Scientific Chemicals
Catalog number191185000
Price (USD)/ Each
135.65
Online exclusive
150.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS20816-12-0
IUPAC Nametetraoxoosmium
Molecular FormulaO4Os
InChI KeyVUVGYHUDAICLFK-UHFFFAOYSA-N
SMILESO=[Os](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Osmium (Os)74.2 to 75.4 %
SolubilityPasses test
Appearance (Color)Yellow
Appearance (Form)Crystals
Non-volatile matter=<0.2 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis, Oxidation reagent
RUO – Research Use Only