Thermo Scientific Chemicals

Aluminum tri-sec-butoxide, 97%, Thermo Scientific Chemicals

Catalog number: 191210025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

Aluminum tri-sec-butoxide, 97%, Thermo Scientific Chemicals

Catalog number: 191210025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 191210025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2269-22-9
IUPAC Name
aluminium(3+) tris(butan-2-olate)
Molecular Formula
C12H27AlO3
InChI Key
WOZZOSDBXABUFO-UHFFFAOYNA-N
SMILES
[Al+3].CCC(C)[O-].CCC(C)[O-].CCC(C)[O-]
Appearance (Color)
Clear to slightly hazy colorless to light yellow
Infrared spectrum
Conforms
Assay
>=96.0 % (ex Al)
Refractive index
1.4380 to 1.4400 (20°C, 589 nm)
Appearance (Form)
Viscous liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text