Thermo Scientific Chemicals

Antimony trifluoride, 98%, Thermo Scientific Chemicals

Catalog number: 191245000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Antimony trifluoride, 98%, Thermo Scientific Chemicals

Catalog number: 191245000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 191245000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7783-56-4
IUPAC Name
antimony(3+) trifluoride
Molecular Formula
F3Sb
InChI Key
GUNJVIDCYZYFGV-UHFFFAOYSA-K
SMILES
[F-].[F-].[F-].[Sb+3]
Titration Iodimetric
>=97.5 %
Appearance (Color)
White to grey
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text