Thermo Scientific Chemicals

Lithium hexafluorophosphate, 98%, pure, Thermo Scientific Chemicals

Catalog number: 191261000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Lithium hexafluorophosphate, 98%, pure, Thermo Scientific Chemicals

Catalog number: 191261000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 191261000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
21324-40-3
IUPAC Name
lithium(1+) hexafluoro-λ⁵-phosphanuide
Molecular Formula
F6LiP
InChI Key
AXPLOJNSKRXQPA-UHFFFAOYSA-N
SMILES
[Li+].F[P-](F)(F)(F)(F)F
Appearance (Form)
Powder
Appearance (Color)
White to off-white
Assay
>=97.5 % (ex Li)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text