Thermo Scientific™

CaptureSelect™ Human Albumin Affinity Matrix

Catalog number: 1912970250
Have Questions?
Thermo Scientific™

CaptureSelect™ Human Albumin Affinity Matrix

Catalog number: 1912970250
Have Questions?
Catalog Number
1912970250
Unit Size
250 mL
Price (USD)
Availability
-
Quantity
-
Catalog Number
Unit Size
5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 L
Price (USD)
Availability
-
Quantity
-
Catalog Number
Unit Size
5 L
Price (USD)
Availability
-
Quantity
-
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1912970250250 mL
--
1912970055 mL
-
19129701010 mL
-
19129705050 mL
-
19129701L1 L
--
19129705L5 L
--
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

CaptureSelect™ Human Albumin Affinity Matrix has been specifically designed for the purification of human albumin and albumin fusion proteins.

Features of this affinity matrix include:

  • Highly selective for human albumin
  • Unique elution characteristics allowing for gentle elution of albumin fusion proteins
  • Excellent scalability
  • Non-animal-derived

The high selectivity and yields obtained using CaptureSelect™ Human Albumin Affinity Matrix enable a robust and efficient purification process with excellent purity obtained in one step. The mild elution conditions make the resin suitable for the purification of albumin fusion proteins, especially those that are pH sensitive.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main Characteristics:

  • Matrix: agarose-based, aldehyde-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect™ HSA affinity ligand
  • Ligand coupling method: aldehyde coupling via NH2 residues of the ligand
  • Binding capacity: ∼15 mg albumin/mL resin
  • Elution conditions (neutral pH): 20 mM Tris, pH 7.4 with 2 M MgCl2 or with 1 M NaCl + 50%(v/v) propylene glycol or with 1 M NaCl + 0.5 M L-arginine
  • Elution conditions (acidic pH): 0.1 M glycine, pH 3.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20%(v/v) ethanol

Specifications

Binding Capacity
∼15 mg albumin/mL resin
Flow Rate
50 to 200 cm/hr. (Up to 2 bar)
Matrix
Agarose-based, aldehyde-activated
Particle Size
65 ± 10 μm
Form
Liquid Suspension
Product Line
CaptureSelect™
Quantity
250 mL
Type
Affinity Matrix
Ligand Type
HSA affinity ligand
Stationary Phase
Anti-HSA
Format
Bottle

Contents & Storage

Store in refrigerator (2° to 8°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text