Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
191304050 | 50 mL |
191304005 | 5 mL |
191304010 | 10 mL |
CaptureSelect IgG3 (Hu) Affinity Matrix can be used for the purification and isolation of IgG3, without cross-reactivity to other immunoglobulins, like other human IgG subclasses, IgA, and IgM, from complex sources like plasma, serum, and cell culture supernatants.