CaptureSelect™ Transferrin Affinity Matrix
CaptureSelect™ Transferrin Affinity Matrix
Thermo Scientific™
CaptureSelect™ Transferrin Affinity Matrix
This is one of a group of Thermo Scientific™ CaptureSelect™ protein purification affinity resins developed for the simple, single-step purification of a variety human plasma and serum proteins.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
19130601010 mL
1913060055 mL
19130605050 mL
3 Options
Catalog number 191306010
Price (USD)/ 10 mL
2,170.00
-
Add to cart
Quantity:
10 mL
Price (USD)/ 10 mL
2,170.00
Add to cart
CaptureSelect™ Transferrin Affinity Matrix
Catalog number191306010
Price (USD)/ 10 mL
2,170.00
-
Add to cart
This is one of a group of CaptureSelect protein purification affinity resins developed for the simple, single-step purification of a variety human plasma and serum proteins. This unique selection of affinity purification resins can purify proteins or other non-antibody biomolecules from either recombinant or native sources with a high degree of affinity and specificity, and are immobilized on a high capacity agarose support.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormLiquid Suspension
Product LineCaptureSelect™
Quantity10 mL
TypeAffinity Matrix
Stationary PhaseAnti-transferrin
Unit Size10 mL
Contents & Storage
Store at 2°C to 8°C.