Chemical Identifiers
CAS6104-58-1
IUPAC Namesodium {4-[(4-ethoxyphenyl)amino]phenyl}bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}-2-methylphenyl)methylium
Molecular FormulaC47H48N3NaO7S2
InChI KeyCMGWKYXJIYYESN-UHFFFAOYSA-M
SMILES[Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2C)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2C)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1
View more