Brilliant Blue G, pure
Brilliant Blue G, pure
Thermo Scientific Chemicals

Brilliant Blue G, pure

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 191485000
Price (USD)/ Each
379.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
446.00 
Save 66.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
379.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
446.00 
Save 66.35 (15%)
Add to cart
Brilliant Blue G, pure
Catalog number191485000
Price (USD)/ Each
379.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
446.00 
Save 66.35 (15%)
-
Add to cart
Chemical Identifiers
CAS6104-58-1
IUPAC Namesodium {4-[(4-ethoxyphenyl)amino]phenyl}bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}-2-methylphenyl)methylium
Molecular FormulaC47H48N3NaO7S2
InChI KeyCMGWKYXJIYYESN-UHFFFAOYSA-M
SMILES[Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2C)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2C)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
UV Lambda max608 to 618 nm (in 50:50 IMS:H2O)
Specific absorbance A (1%/1cm)>=350 (at lambda max.)
Appearance (Form)Powder
HPLC>=80 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only