Thermo Scientific Chemicals

Brilliant Blue R, pure

Catalog number: 191491000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Brilliant Blue R, pure

Catalog number: 191491000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 191491000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6104-59-2
IUPAC Name
sodium {4-[(4-ethoxyphenyl)amino]phenyl}bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)methylium
Molecular Formula
C45H44N3NaO7S2
InChI Key
HAEGGLCXLFWTBE-UHFFFAOYSA-M
SMILES
[Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1
Appearance (Color)
Bordeaux to dark purple-brown
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
Absorption Ratio P-15/P+15
1 to 1.2
UV Lambda max
588 to 596 nm (in 50:50 ethanol:water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text