Thermo Scientific Chemicals

2',7'-Dichlorofluorescein, pure

Catalog number: 191531000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2',7'-Dichlorofluorescein, pure

Catalog number: 191531000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 191531000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
76-54-0
IUPAC Name
2',7'-dichloro-3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular Formula
C20H10Cl2O5
InChI Key
VFNKZQNIXUFLBC-UHFFFAOYSA-N
SMILES
OC1=CC2=C(C=C1Cl)C1(OC(=O)C3=CC=CC=C13)C1=CC(Cl)=C(O)C=C1O2
Specific absorbance A (1%/1cm)
>=2000 (at lambda max.)
Appearance (Color)
Light orange to red-brown
Infrared spectrum
Conforms
UV Lambda max
501 to 503 nm (in 0.1 N NaOH)
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text