Thermo Scientific Chemicals

Fast Blue BB, 98%, pure

Catalog number: 191570250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Fast Blue BB, 98%, pure

Catalog number: 191570250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 191570250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
120-00-3
IUPAC Name
N-(4-amino-2,5-diethoxyphenyl)benzamide
Molecular Formula
C17H20N2O3
InChI Key
CNXZLZNEIYFZGU-UHFFFAOYSA-N
SMILES
CCOC1=CC(NC(=O)C2=CC=CC=C2)=C(OCC)C=C1N
Infrared spectrum
Conforms
Appearance (Color)
Grey to brown
HPLC
>=97.5 %
Appearance (Form)
Powder and/or chunks
UV spectrum
Complies

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text