Thermo Scientific Chemicals

Fluorescamine, pure, Thermo Scientific Chemicals

Catalog number: 191670025
2.5 g, Glass bottle, Each
Thermo Scientific Chemicals

Fluorescamine, pure, Thermo Scientific Chemicals

Catalog number: 191670025
2.5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 191670025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
38183-12-9
IUPAC Name
4'-phenyl-3H,3'H-spiro[2-benzofuran-1,2'-furan]-3,3'-dione
Molecular Formula
C17H10O4
InChI Key
ZFKJVJIDPQDDFY-UHFFFAOYNA-N
SMILES
O=C1OC2(OC=C(C2=O)C2=CC=CC=C2)C2=CC=CC=C12
Specific extinction E (1%/1cm)
>=900 (235 nm, ether)
Infrared spectrum
Conforms
Suitability
fluorescence ind. for prim. amines and amino acids
Appearance (Form)
Powder
Melting point
155°C to 157°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text