Chemical Identifiers
CAS2869-83-2
IUPAC Name3-(diethylamino)-7-{2-[4-(dimethylamino)phenyl]diazen-1-yl}-5-phenyl-5λ⁵-phenazin-5-ylium chloride
Molecular FormulaC30H31ClN6
InChI KeyXXACTDWGHQXLGW-UHFFFAOYSA-M
SMILES[Cl-].CCN(CC)C1=CC=C2N=C3C=CC(=CC3=[N+](C3=CC=CC=C3)C2=C1)N=NC1=CC=C(C=C1)N(C)C
View more