Chemical Identifiers
CAS12790-48-6
IUPAC Namebis(λ⁵-ruthenium(5+)) tetrahydrate ruthenium tetradecaazanide hexachloride dioxidandiide
Molecular FormulaCl6H36N14O6Ru3
InChI KeyIMVKVBRYNZXYAP-UHFFFAOYSA-H
SMILES[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].O.O.O.O.[O--].[O--].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru].[Ru+5].[Ru+5]
View more